missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Cypionate, TRC
Specifications
Specifications
| Chemical Name or Material | Testosterone Cypionate |
| CAS | 58-20-8 |
| Molecular Formula | C27 H40 O3 |
| Quantity | 1 g |
| Synonym | Androst-4-en-3-one','17-(3-cyclopentyl-1-oxopropoxy)-','(17β)-, Testosterone','cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid','ester with testosterone (8CI), 17β-Hydroxyandrost-4-en-3-one cyclopentanepropionate, 17β-Hydroxyandrost-4-en-3-one cyclopentylpropionate, DepAndro 100, DepAndro 200, Depo, Depo-testosterone, Deposteron, Depotest, Depovirin, NSC 9157, Pertestis, Testex Leo, Testodrin prolongatum, Testosterone 17β-cyclopentanepropionate, Testosterone 17β-cyclopentylpropionate, Testosterone 17β-cypionate, Testosterone cyclopentylpropionate, Testosterone cypionate, Testosterone','3-cyclopentanepropionate, Virilon |
| InChI Formula | InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5 |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate |
| Molecular Weight (g/mol) | 412.6 |
| Formula Weight | 412.2977 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?