Learn More
Testosterone 17beta-Carboxylic Acid, TRC
Product Code. 30427316
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427316 2.5 g 2.5g
30412127 100 mg 100mg
30420167 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30427316 Supplier Toronto Research Chemicals Supplier No. TRCT1550702.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 302-97-6
Molecular Formula C20 H28 O3
Molecular Weight (g/mol) 316.43
Synonym (17β)-3-Oxo-androst-4-ene-17-carboxylic Acid, Androst-4-ene-17β-carboxylic acid, 3-oxo- (6CI,7CI,8CI), 17β-Carboxyandrost-4-en-3-one, 3-Carbonyl-4-androstene-17β-carboxylic acid, 3-Oxoandrost-4-ene-17β-carboxylic acid, AD-Acid, AL 3793, CC 14837, Etienic acid, L 552.803, NSC 226121, SKF 106224, Testosterone-17β-carboxylic acid, 4-Androsten-3-one-17β-carboxylic Acid, 3-Oxo-4-androstene-17β-carboxylic Acid
IUPAC Name (8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-17-carboxylic acid
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)O
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C20H28O3/c1-19-9-7-13(21)11-12(19)3-4-14-15-5-6-17(18(22)23)20(15,2)10-8-16(14)19/h11,14-17H,3-10H2,1-2H3,(H,22,23)/t14-,15-,16-,17+,19-,20-/m0/s1
Formula Weight 316.2038
Chemical Name or Material 4-Androsten-3-one-17beta-carboxylic Acid