Learn More
Testosterone-16,16,17-d3 17-Decanoate, TRC
Product Code. 30428053
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428053 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30428053 Supplier Toronto Research Chemicals Supplier No. TRCT1551221MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Testosterone-16,16,17-d3 17-Decanoate
CAS 876054-57-8
Molecular Formula C29 2H3 H43 O3
Quantity 1 mg
Synonym Androst-4-en-3-one-16,16,17-d3','17-[(1-oxodecyl)oxy]-','(17β)- (9CI), Testosterone-16,16,17-d3 17-Decanoate
InChI Formula InChI=1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1/i15D2,26D
SMILES [2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])OC(=O)CCCCCCCCC
IUPAC Name [(8R,9S,10R,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] decanoate
Molecular Weight (g/mol) 445.69
Formula Weight 445.3635