Learn More
Cole-Parmer™ Testing Sieve, Brass
Product Code. 15872145 Shop All Cole-Parmer Products
Change view
Click to view available options
Pore Size:
2 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
15872145 2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15872145 Supplier Cole-Parmer™ Supplier No. 5998101
  • ONLINE EXCLUSIVE PRICE
    109.00€
    84.92€ / Pack of 1
    Save 24.08€
    22% Off
Estimated Shipment: 19-11-2026
Serialized sieve with a test sieve certificate of compliance.
  • Brass frame, brass wire.
  • Test sieve certificate.
  • Equivalent to ASTM E11-09 compliance sieve.
Specifications
Pore Size 2 mm
Material Brass
Height (English) 1 in.
U.S. Standard No. 10
Diameter (Metric) Outer 8 in.
Screen Size 10 Mesh
Mesh Material Brass
Sieve Number 10