Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Conical Bottom
Product Code. 11943571
Change view
Click to view available options
Diameter (Metric):
17 mm
Length (Metric):
115 mm
120 mm
Closure Type:
Hexagonal
Not Included
Stopper
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Closure Type Unit Size
11943571 17 mm 120 mm Not Included Pack of 10
10176684 17 mm 115 mm Stopper Pack of 10
11923571 17 mm 115 mm Not Included Pack of 10
10667780 17 mm 120 mm Stopper Pack of 10
10677780 17 mm 120 mm Hexagonal Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11943571 Supplier Lenz Laborglasintrumente Supplier No. 05705016
  • ONLINE EXCLUSIVE PRICE
    131.00€
    81.84€ / Pack of 10
    Save 49.16€
    37% Off
Estimated Shipment: 09-11-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With conical bottom
  • With socket NS 14/23
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing
Specifications
Closure Material Polyethylene
Length (Metric) 120 mm
Material Borosilicate Glass (3.3)
Diameter (Metric) 17 mm
Closure Type Not Included
Bottom Shape Conical