Learn More
Velp Scientifica™ Test Tubes for Digestion Units
Product Code. p-8012702 Shop All Velp Products
Click to view available options
For Use With (Equipment):
DK6/48 and UDK series
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
UDK Series distillation units
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
Unit Size:
Each
Pack of 3
Pack of 6
Product Code. 11725654
Brand: Velp Scientifica™ A00000146
Includes
6 Test Tubes/Box
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available in different counts
  • Available in different sizes and diameters
Specifications
Product Type Test Tubes
Capacity (Metric) 100 mL
Diameter (Metric) Outer 26 mm
For Use With (Equipment) Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
Quantity 6 Tubes/ Box