Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Product Code. 10590823
Click to view available options
Material:
Polypropylene
Stainless Steel
Holds:
21 Test Tubes, 30/31 mm dia.
90 Microliter Tubes, 11/12 mm dia.
90 Test Tubes, 12/13 mm dia.
Unit Size:
Each
Product Code. 10590823
Brand: Julabo™ 8970382
Specifications
Description Test tube rack
For Use With (Equipment) SW22, SW23, TW12, TW20, TW8 shaking water bath
Holds 90 Microliter Tubes, 11/12 mm dia.