Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Product Code. 11476358 Shop All Thermo Scientific Products
Click to view available options
Color:
Blue
Green
Orange
Red
White
Holds:
Tubes (1.5 mL)
Tubes (10 to 13 mm)
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Tubes (17 to 20 mm)
Tubes (21 to 25 mm)
Tubes (26 to 30 mm)
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Array:
3 x 3
3 x 8 array
4 x 10
4 x 4
4 x 5
4 x 6
6 x 12
6 x 6
8 x 12
Unit Size:
Each
Product Code. 11476358
Brand: Thermo Scientific™ 30185
Only null left
Add to basket
Add to basket
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Tube Rack
Description White; 17 to 20 mm; 4 x 5 array
Array 4 x 5
Color White
Holds Tubes (17 to 20 mm)
Positions 20
For Use With (Equipment) MaxQ™ Shaker Platform