Learn More
Julabo™ Test Tube Rack
Product Code. p-7160845
Change view
Click to view available options
No. of Positions:
21
30
42
60
90
Unit Size:
Each
Product Code. No. of Positions unitSize
15470632 21 Each
15480632 30 Each
15400642 42 Each
15430632 60 Each
15450632 90 Each
5 options
Product Code. 15470632
Brand: Julabo™ 9970303
Test tube rack made of high grade plastic or stainless steel
Specifications
Description Test tube racks for water baths
Temperature Range Up to +100°C
Material High grade plastic
No. of Positions 21
Size 30 mm dia.