Learn More
OHAUS™ Test Tube Rack
Shop All Ohaus Products
Click to view available options
Positions:
12
17
21
24
33
35
40
48
60
63
90
Holds:
Tubes (10 to 13 mm)
Tubes (10 to 14 mm)
Tubes (14 to 16 mm)
Tubes (15 mL)
Tubes (16 mm)
Tubes (16 to 20 mm)
Tubes (18 to 20 mm)
Tubes (20 mm)
Tubes (21 to 25 mm)
Tubes (22 to 25 mm)
Tubes (25 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
Description
- Constructed of stainless steel
- For use with tubes of various sizes
Specifications
Specifications
| Product Type | Test Tube Rack |
| Description | Test Tube Rack 10-13 mm Diameter |
| Array | 7 x 9 |
| Autoclavable | Autoclavable |
| Color | Blue |
| Holds | Tubes (10 to 13 mm) |
| Material | PVC-coated Steel |
| Positions | 63 |
| Shape | Rectangular |
| Shelves | 1 |
| Show More |