Learn More
RETSCH Test Sieve Rack
Product Code. 10278610 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Vibratory Sieve Shaker AS 200
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10278610 Vibratory Sieve Shaker AS 200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10278610 Supplier RETSCH Supplier No. 32.012.0001
  • ONLINE EXCLUSIVE PRICE
    377.00€
    355.81€ / Pack of 1
    Save 21.19€
    5% Off
Estimated Shipment: 07-12-2026
Specifications
Product Type Test Sieve Rack
Diameter (Metric) 200 mm
For Use With (Equipment) Vibratory Sieve Shaker AS 200
Packaging Quantity 1 Ea.