missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Total Chlorine Reagents
Click to view available options
Range:
0 to .35 mg/L
0 to 1 mg/L
0 to 15 mg/L
0 to 2.5 mg/L
0 to 5 mg/L
Unit Size:
100g
Pack of 100
Description
- Reagents for the colorimetric determination of nitrite
- There are enough reagents for 100 tests to be used with a compatible benchtop or portable photometer
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- Marked with expiration date and lot number for traceability
- High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
Specifications
Specifications
| Quantity | 100 Pack |
| Range | 0 to .35 mg/L |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : potassium disulphate, chromotropic aciddisodium salt dihydrate, trans-diamino-1,2-cyclohexane-N,N,N
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction