Learn More
Hanna Instruments™ Total Chlorine Reagents
Product Code. 11867843
Click to view available options
Range:
0 to .35 mg/L
0 to 1 mg/L
0 to 15 mg/L
0 to 2.5 mg/L
0 to 5 mg/L
Unit Size:
100g
Pack of 100
Product Code. 11867843
Brand: Hanna Instruments™ HI9370701
Prepared with high purity chemicals, these high quality reagents are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
  • Reagents for the colorimetric determination of nitrite
  • There are enough reagents for 100 tests to be used with a compatible benchtop or portable photometer
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
  • Marked with expiration date and lot number for traceability
  • High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
Specifications
Quantity 100 Pack
Range 0 to .35 mg/L
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : potassium disulphate, chromotropic aciddisodium salt dihydrate, trans-diamino-1,2-cyclohexane-N,N,N