missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies TeSR™-E8™ Kit for hESC/hiPSC Maintenance

Product Code. 15889944
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15889944

Brand: STEMCELL Technologies 05990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Feeder-free, animal component-free culture medium for maintenance of human ES and iPS cells

  • TeSR™-E8™ is a feeder-free, animal component-free culture medium for human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells
  • It is based on the E8 formulation developed by the laboratory of Dr
  • James Thomson (University of Wisconsin-Madison), the lead research group behind the design of mTeSR™1, the most widely published feeder-free culture medium for pluripotent stem cells
  • Like mTeSR™1, TeSR™-E8™ medium is made with the highest level of quality and care
  • It contains only the essential components required for maintenance of ES and iPS cells, providing a simpler medium for the culture of pluripotent stem cells
  • TeSR™-E8™ may be used with either Corning™ Matrigel™ hESC-Qualified Matrix (Catalog #07181) or, for a completely defined xeno-free system, Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics) as the culture matrix

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Expansion, Maintenance
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.