missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies TeSR™-E5

Product Code. 15887035
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887035 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887035 Supplier STEMCELL Technologies Supplier No. 05916

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined, serum-free, xeno-free medium for pluripotent stem cells

  • TeSR™-E5 is a defined, serum- and xeno-free medium that is based on the formulation of TeSR™-E8™, but does not contain transforming growth factor β (TGF-β), basic fibroblast growth factor (bFGF) or insulin
  • It may be used as a basal medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells, or other applications where removal of the above cytokines and insulin is desirable

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Characterization, Differentiation
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.