Learn More
Terumo™ Hypodermic Needles
Product Code. p-184970
Click to view available options
Needle Gauge:
20 G
22 G
Length (English) Needle:
1 in.
1.5 in.
Needle Point Style:
Ultra-thin Wall
Unit Size:
Pack of 100
Product Code. 11587312
Brand: Terumo™ 2NN2238R
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides ultra sharp needles, which translate into greater comfort for the patient.
  • Double bevel design
  • Precision manufactured ultrathin wall
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 22 G
Latex Free Yes
Length (English) Needle 1.5 in.
Product Type Double Bevel Design
Needle Point Style Ultra-thin Wall