Learn More
tert-Butyl ethyl ether, 99%
Product Code. 11450018
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11450018
Brand: Thermo Scientific Alfa Aesar B22945.14
Request Bulk or Custom Format
CAS: 637-92-3 | C6H14O | 102.18 g/mol
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C
Specifications
Melting Point -94°C
Density 0.75
Boiling Point 72°C to 73°C
Flash Point −19°C (−2°F)
Refractive Index 1.375
Quantity 25 g
UN Number UN1179
Beilstein 1731469
Merck Index 14,3774
Solubility Information Miscible with alcohol,ethyl ether. Slightly miscible with water.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material tert-Butyl ethyl ether