missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Acetate-d12, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Acetic Acid tert-Butyl Ester D12 |
| CAS | 358731-05-2 |
| Molecular Formula | C6 D12 O2 |
| Quantity | 0.25 g |
| Synonym | Acetic-d3 acid, 1,1-di(methyl-d3)ethyl-2,2,2-d3 ester (9 CI), Acetic acid tert-butyl ester D12, tert-Butyl acetate-d12, tert-Butyl-d3 acetate-d3, Acetic-d3 acid 1,1-di(methyl-d3)ethyl-2,2,2-d3 ester |
| InChI Formula | InChI=1 S/C6H12O2/c1-5(7)8-6(2,3)4/h1-4H3/i1D3,2D3,3D3,4D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)OC(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | [1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl] 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 128.232 |
| Formula Weight | 128.159 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction