Learn More
tert-Butyl 8-Hydroxyoctanoate, TRC
Product Code. 30343661
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343661 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30343661 Supplier Toronto Research Chemicals Supplier No. TRCB6932251G
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material tert-Butyl 8-Hydroxyoctanoate
CAS 401901-41-5
Molecular Formula C12 H24 O3
Quantity 1 g
Synonym 8-Hydroxyoctanoic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C12H24O3/c1-12(2,3)15-11(14)9-7-5-4-6-8-10-13/h13H,4-10H2,1-3H3
SMILES CC(C)(C)OC(=O)CCCCCCCO
IUPAC Name tert-butyl 8-hydroxyoctanoate
Molecular Weight (g/mol) 216.32
Formula Weight 216.1725
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.