Learn More
tert-Butyl-7-[4-(4-fluorophenyl)-6-isopropyl-2-mesylaminopyrimidin-5-yl]-(3R,5S)-isopropylidine-(E)-6-heptenoate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl-7-[4-(4-fluorophenyl)-6-isopropyl-2-mesylaminopyrimidin-5-yl]-(3R,5S)-isopropylidine-(E)-6-heptenoate |
| CAS | 371775-73-4 |
| Molecular Formula | C28H38FN3O6S |
| Quantity | 10 mg |
| Synonym | (4R,6S)-6-[(1E)-2-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[(methylsulfonyl)amino]-5-pyrimidinyl]ethenyl]-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C28H38FN3O6S/c1-17(2)24-22(25(18-9-11-19(29)12-10-18)31-26(30-24)32-39(8,34)35)14-13-20-15-21(37-28(6,7)36-20)16-23(33)38-27(3,4)5/h9-14,17,20-21H,15-16H2,1-8H3,(H,30,31,32)/b14-13+/t20-,21-/m1/s1 |
| SMILES | CC(C)c1nc(NS(=O)(=O)C)nc(c1\C=C\[C@@H]2C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O2)c3ccc(F)cc3 |
| IUPAC Name | tert-butyl 2-[(4R,6S)-6-[(E)-2-[4-(4-fluorophenyl)-2-(methanesulfonamido)-6-propan-2-ylpyrimidin-5-yl]ethenyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate |
| Molecular Weight (g/mol) | 563.68 |
| Formula Weight | 563.2465 |