Learn More
tert-Butyl 6-Hydroxyhexanoate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl 6-Hydroxyhexanoate |
| CAS | 73839-20-0 |
| Molecular Formula | C10 H20 O3 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C10H20O3/c1-10(2,3)13-9(12)7-5-4-6-8-11/h11H,4-8H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)CCCCCO |
| IUPAC Name | tert-butyl 6-hydroxyhexanoate |
| Molecular Weight (g/mol) | 188.26 |
| Formula Weight | 188.1412 |