Learn More
tert-Butyl 6-Chloro-5(S)-hydroxy-3-oxohexanoate, TRC
Product Code. 30240827
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240827 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30240827 Supplier Toronto Research Chemicals Supplier No. TRCB69088550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154026-92-3
Molecular Formula C10 H17 Cl O4
Molecular Weight (g/mol) 236.69
Synonym Hexanoic acid, 6-chloro-5-hydroxy-3-oxo-, 1,1-dimethylethyl ester, (5S)-, Hexanoic acid, 6-chloro-5-hydroxy-3-oxo-, 1,1-dimethylethyl ester, (S)-, (5S)-6-Chloro-5-hydroxy-3-oxohexanoic acid tert-butyl ester, (5S)-6-Chloro-5-hydroxy-3-oxohexanoic acid tert-butyl ester, Tert-Butyl 6-chloro-5(S)-hydroxy-3-oxohexanoate
IUPAC Name tert-butyl (5S)-6-chloro-5-hydroxy-3-oxohexanoate
SMILES CC(C)(C)OC(=O)CC(=O)C[C@H](O)CCl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H17ClO4/c1-10(2,3)15-9(14)5-7(12)4-8(13)6-11/h8,13H,4-6H2,1-3H3/t8-/m0/s1
Formula Weight 236.0815
Chemical Name or Material tert-Butyl 6-Chloro-5(S)-hydroxy-3-oxohexanoate