Learn More
tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate, ≥97%, Thermo Scientific™
Product Code. p-3174933
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. 10632065
Brand: Thermo Scientific Maybridge CC43909CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 857284-20-9
Molecular Formula C15H23N3O3
Molecular Weight (g/mol) 293.367
MDL Number MFCD08060526
InChI Key QKCHWJYIPFBVRD-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl pyrid-2-yl piperazine-1-carboxylate, tert-butyl 4-5-hydroxymethyl pyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-5-hydroxymethyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-hydroxymethyl-2-pyridinyl-,1,1-dimethylethyl ester
PubChem CID 7537576
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)CO
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 293.36
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate