Learn More
tert-Butoxybis(dimethylamino)methane, tech. 90%
Product Code. 11367469
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11367469
Brand: Thermo Scientific Alfa Aesar H27861.06
Request Bulk or Custom Format
CAS: 5815-08-7 | C9H24N2O | 176.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5815-08-7
Molecular Formula C9H24N2O
Molecular Weight (g/mol) 176.30
MDL Number MFCD00042858
InChI Key HXRAMSFGUAOAJR-UHFFFAOYSA-P
Synonym tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane
PubChem CID 79885
IUPAC Name N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
SMILES CC(C)(C)OC(N(C)C)N(C)C
Specifications
Density 0.844
Boiling Point 50°C to 55°C (15 mmHg)
Flash Point 41°C (106°F)
Linear Formula (CH3)3COCH[N(CH3)2]2
Refractive Index 1.422
Quantity 5 g
UN Number UN1993
Formula Weight 174.3
Percent Purity 90%
Grade Technical
Chemical Name or Material tert-Butoxybis(dimethylamino)methane