missing translation for 'onlineSavingsMsg'
Learn More

Melford Terrific Broth, Capsules

Product Code. 30018824
Click to view available options
Quantity:
600 G
1.2 kg
5 kg
Unit Size:
1.2kg
5kg
600g
This item is not returnable. View return policy

Product Code. 30018824

Brand: Melford T150505000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Terrific Broth formulation packaged in easy to use 24 gram capsules.

Terrific Broth formulation packaged in easy to use 24 gram capsules. Formulation from powder.

Add 1 capsule per 500ml of water. Heat and stir until capsules are completely dissolved. Add 2ml of glycerol to mixture. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 12 g/L

Yeast Extract: 24 g/L

Dipotassium Phosphate: 9.4 g/L

Potassium Phosphate: 2.2 g/L

Specifications

Quantity 5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.