Learn More
3M™ Nomad™ Terra Medium Traffic Entrance Matting 6050
Click to view available options
Dimensions (L x W):
152 x 91.4 cm
610 x 91.4 cm
180 x 122 cm
610 x 122 cm
Color:
Black
Blue
Brown
Gray
Red
Unit Size:
1 roll
Each
Description
- First step mat for efficient dirt removal
- Suitable for medium traffic volumes
- Collects and prevents dirt from being tracked into the building
- Holds and hides dirt & sand, so the surface stays neat and clean
- Foam backing reduces creeping and, together with the coils, provides additional cushioning for increased comfort
- Easy installation
Specifications
Specifications
| Dimensions (L x W) | 610 x 122 cm |
| For Use With (Application) | Medium traffic areas |
| Thickness (Metric) | 11 mm |
| Color | Black |
| Length (Metric) | 610 cm |
| Material | Vinyl |
| Type | Entrance Mat |
| Width (Metric) | 122 cm |