Learn More
3M™ Nomad™ Terra Medium Traffic Entrance Matting 6050
Product Code. 17168720
Click to view available options
Dimensions (L x W):
152 x 91.4 cm
610 x 91.4 cm
180 x 122 cm
610 x 122 cm
Color:
Black
Blue
Brown
Gray
Red
Unit Size:
1 roll
Each
Product Code. 17168720
Brand: 3M™ Nomad™ 7100116695
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This backed vinyl matting is suitable for indoor, medium traffic applications. Resilient vinyl loop construction traps, holds, and hides dirt/sand so surfaces stay neat and clean.
  • First step mat for efficient dirt removal
  • Suitable for medium traffic volumes
  • Collects and prevents dirt from being tracked into the building
  • Holds and hides dirt & sand, so the surface stays neat and clean
  • Foam backing reduces creeping and, together with the coils, provides additional cushioning for increased comfort
  • Easy installation
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 610 x 122 cm
For Use With (Application) Medium traffic areas
Thickness (Metric) 11 mm
Color Red
Length (Metric) 610 cm
Material Vinyl
Type Entrance Mat
Width (Metric) 122 cm