missing translation for 'onlineSavingsMsg'
Learn More

Tergitol NP-10 (Technical Grade), TRC

Product Code. 30456991
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456991 100 mL 100mL
30454482 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456991 Supplier Toronto Research Chemicals Supplier No. TRCT548815100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127087-87-0
SMILES O=C(COCC([CPG-Icaa])=O)OC(COC(C1=CC=C(OC)C=C1)(C2=CC=CC=C2)C3=CC=C(OC)C=C3)COC4=C5CCCN6C5=C(CCC6)C=C4/N=N/C7=C(OC)C=C(/N=N/C8=CC=C([N+]([O-])=O)C=C8)C(OC)=C7

Specifications

Quantity 100 mL
Chemical Name or Material Tergitol NP-10 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.