Learn More
(±)-Terconazole-d4 (piperazine-2,2,6,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Terconazole-D4 (piperazine-2,2,6,6-D4) |
| CAS | 1398065-50-3 |
| Molecular Formula | C26 D4 H27 Cl2 N5 O3 |
| Quantity | 0.01 g |
| Synonym | (±)-Terconazole-d4 (piperazine-2,2,6,6-d4), Terconazole-D4 (piperazine-2,2,6,6-D4), Terconazole-d4, rel-1-[4-[[(2 R,4 S)-2-(2,4-Dichlorophenyl)-2-(1 H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-(1-methylethyl)piperazine-2,2,6,6-d4 |
| InChI Formula | InChI=1 S/C26H31Cl2N5O3/c1-19(2)31-9-11-32(12-10-31)21-4-6-22(7-5-21)34-14-23-15-35-26(36-23,16-33-18-29-17-30-33)24-8-3-20(27)13-25(24)28/h3-8,13,17-19,23 H,9-12,14-16H2,1-2H3/t23-,26-/m0/s1/i11D2,12D2 |
| SMILES | [2 H]C1([2 H])CN(CC([2 H])([2 H])N1c2ccc(OC[C@H]3 CO[C@@](Cn4cncn4)(O3)c5ccc(Cl)cc5Cl)cc2)C(C)C |
| IUPAC Name | 2,2,6,6-tetradeuterio-1-[4-[[(2 R,4 S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-propan-2-ylpiperazine |
| Molecular Weight (g/mol) | 536.487 |
| Formula Weight | 535.206 g/mol |
| Show More |