Learn More
Tocris Bioscience™ Terbinafine hydrochloride
Description
Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.
Specifications
Specifications
| CAS | 78628-80-5 |
| Synonym | N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride |
| Quantity | 50 mg |
| Molecular Formula | C21H25N.HCl |
| Formula Weight | 327.89 |
| Purity | 98% |