Learn More
Terbinafine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Terbinafine-d3 HCl (N-methyl-d3) |
| CAS | 1310012-15-7 |
| Molecular Formula | C21H22D3N.HCl |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C21H25N.ClH/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19;/h5-7,9-14 H,16-17H2,1-4H3;1 H/b9-5+;/i4D3; |
| SMILES | CC(C)(C)C#C/C=C/CN(C([2 H])([2 H])[2 H])CC1=CC=CC2=CC=CC=C21.Cl |
| IUPAC Name | (E)-6,6-dimethyl-N-(methyl-d3)-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride |
| Molecular Weight (g/mol) | 330.91 |
| Formula Weight | 330.2 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |