Learn More
Teprenone, TRC
Specifications
Specifications
| Chemical Name or Material | Teprenone |
| CAS | 6809-52-5 |
| Molecular Formula | C23 H38 O |
| Quantity | 25 mg |
| Synonym | 5,9,13,17-Nonadecatetraen-2-one','6,10,14,18-tetramethyl- (6CI','7CI','8CI','9CI','ACI), 6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one (ACI), E 0671, E 36U31, Geranylgeranylacetone, GGA, Selbex, Teprenone |
| InChI Formula | InChI=1S/C23H38O/c1-19(2)11-7-12-20(3)13-8-14-21(4)15-9-16-22(5)17-10-18-23(6)24/h11,13,15,17H,7-10,12,14,16,18H2,1-6H3/b20-13+,21-15+,22-17+ |
| SMILES | CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CCC(=O)C)\C)\C)\C)C |
| IUPAC Name | (5E,9E,13E)-6,10,14,18-tetramethylnonadeca-5,9,13,17-tetraen-2-one |
| Molecular Weight (g/mol) | 330.55 |
| Formula Weight | 330.2923 |