missing translation for 'onlineSavingsMsg'
Learn More

Tenovin 1, TRC

Product Code. 30409050
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409050 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409050 Supplier Toronto Research Chemicals Supplier No. TRCT01847010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tenovin 1
CAS 380315-80-0
Molecular Formula C20H23N3O2S
Quantity 10 mg
Synonym N-[[[4-(Acetylamino)phenyl]amino]thioxomethyl]-4-(1,1-dimethylethyl)benzamide, N-((4-Acetamidophenyl)carbamothioyl)-4-(tert-butyl)benzamide
InChI Formula InChI=1S/C20H23N3O2S/c1-13(24)21-16-9-11-17(12-10-16)22-19(26)23-18(25)14-5-7-15(8-6-14)20(2,3)4/h5-12H,1-4H3,(H,21,24)(H2,22,23,25,26)
SMILES CC(=O)Nc1ccc(NC(=S)NC(=O)c2ccc(cc2)C(C)(C)C)cc1
IUPAC Name N-[(4-acetamidophenyl)carbamothioyl]-4-tert-butylbenzamide
Molecular Weight (g/mol) 369.48
Formula Weight 369.1511
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.