Learn More
LABPLAS Temporary Closure Clips
Product Code. 17960145
Click to view available options
For Use With (Equipment):
3500 Series Bags
400 Series Bags
80 Series Bags
Unit Size:
Each
Product Code. 17960145
Brand: LABPLAS SA24060
Only null left
Add to basket
Add to basket
Temporary closure clips to close bags that do not have a closure system.
  • Simple and easy to use
  • Allows you to quickly close the bag and re-open it if needed
  • Available in multiple sizes to suit the size of the bag.
Specifications
Autoclavable Not Autoclavable
Type Clamp
For Use With (Equipment) 400 Series Bags