Learn More
TFA Dostmann™ Temperature Transmitter
Product Code. 11766606
Change view
Click to view available options
Outputs:
-40°C to +160°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Outputs Unit Size
11766606 -40°C to +160°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11766606 Supplier TFA Dostmann™ Supplier No. 30.3120.90
  • ONLINE EXCLUSIVE PRICE
    16.85€
    12.59€ / Pack of 1
    Save 4.26€
    25% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Requires 2 x 1.5V AAA batteries.
  • Display with temperature
  • Range: 100 m in open field
  • Also for temperature monitoring in remote rooms
Specifications
Product Type Temperature Transmitter
Material Plastic
Outputs -40°C to +160°C