missing translation for 'onlineSavingsMsg'
Learn More

BOLA Temperature Probes Lemo (PT 100)

Product Code. 10592552
Change view
Click to view available options
Length (Metric) Overall:
100 mm
200 mm
300 mm
500 mm
600 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Overall unitSize
10592552 500 mm Each
10267021 600 mm Each
15147166 100 mm Each
15530633 200 mm Each
15416124 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10592552 Supplier BOLA Supplier No. P179020
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Temperature Probes Lemo (PT 100) PTFE/Stainless Steel, usable length: PFA-coated connector cable: 1500 mm, with LEMO connector

  • One measuring sensor in a PTFE-encapsulated stainless steel tube (1.471). Temperature probe 8 mm, tip 6 mm, collar ring 12 mm. With white PTFE-coated cable (length: 1.5 m) and coupling (type Lemo™ socket size 1, 4-wire-system).
  • Typical response times:
  • T 50: 7 - 12 s, T 90:14 - 16 s

Specifications

Certifications/Compliance FDA
Disposable Reusable
Length (Metric) Cable 560 mm
Length (Metric) Overall 500 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.