missing translation for 'onlineSavingsMsg'
Learn More

Promed Bioscience Telocollagen, Porcine Type I (Acidic extraction)

Product Code. 17592835
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17592835 10 mg 10mg
17502845 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17592835 Supplier Promed Bioscience Supplier No. PRO200210

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Lyophilized, highly purified Type I collagen (telocollagen), extracted via acid-based method, suitable for 3D cell cultures and tissue engineering applications

  • Lyophilized, highly purified Type I collagen (telocollagen), extracted via acid-based method
  • Telocollagen retains the fibrillar structure of the native molecule and preserves the superior physical and mechanical properties of the collagen found in tissues
  • Suitable as a coating for cell culture labware, for 2D and 3D cell cultures (e.g. higher stiffness scaffolds) and for tissue engineering applications

Specifications

Formulation Lyophilized Fibrous Powder
Purification Method Dialysis
Quantity 10 mg
Content And Storage -20°C to 8°C
Endotoxin Level <1.0 EU/mL
Purity or Quality Grade >95% (by SDS/Western blotting)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.