Learn More
Aalborg™ Model F PTFE/PFA Flowmeter
Product Code. 11787759
Click to view available options
Waterflow Rate:
1000 mL/min.
250 mL/min.
Unit Size:
Each
Product Code. 11787759
Brand: Aalborg™ F6CF0501TF
Only null left
Add to basket
Add to basket
Aalborg™ Model F PTFE/PFA Flowmeter is excellent for high-purity applications or use with corrosive liquids.
  • Chemically inert wetted components
  • Removable protective shield
  • Individually leak tested
  • Vertical in-line
  • Valve at outlet
Specifications
Type Flowmeter
Accuracy ±5%
Pressure Rating 6.7 bar
Connections 1/4 in. NPT (M)
For Use With (Application) Ultra-pure or corrosive liquids
Scale Linear and direct
Temperature (Metric) 121°C
Valve Teflon
Waterflow Rate 1000 mL/min.