missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Teepol Nutrient Pad Set

Product Code. 15952490 Shop All Sartorius Products
Click to view available options
:
10 bags of 10 dishes
This item is not returnable. View return policy

Product Code. 15952490

Brand: Sartorius Stedim Biotech™ 1406750RDN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes dehydrated media pads in petri dishes , pre-sterilized, along with suitable membrane filters for economical, time-saving microbiological quality control.

  • Nutrient pads with gridded membranes
  • For determination of E. coli and coliforms
  • Sterile packaged in petri dishes and individually sealed in bags for longest shelf life.
  • Trouble-free storage up to 24 months
  • Compliance with international standards and references
  • Validation guide available
  • Highly versatile - multiple applications
  • Economical by time- and cost-savings
  • Easy handling
TRUSTED_SUSTAINABILITY

Specifications

Color White
Media Type Teepol/Lauryl Sulfate
Sterility Sterile
Target Organism Class E. coli, Coliforms
Grid Color Green
Packaging Type 10 bags of 10 dishes
Pore Size 0.45μm
Quantity 100/Pk.
Diameter (Metric) 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.