Learn More
Juchheim Laborgeraete™ Teclu Safety Burner
Click to view available options
Gas Type:
Natural Gas
Propane
Height (Metric):
150 mm
200 mm
230 mm
Unit Size:
Each
Description
- For operation with option of natural gas or propane gas
- With needle valve
- With DVGW certification
- With air control screen
- Particularly good stand due to heavy, high-quality material
Specifications
Specifications
| Product Type | Bunsen Burner |
| Gas Type | Natural Gas |
| Material | Brass |
| Description | Gas consumption approx. 116 L/h (0.0813 kg/h), Heating capacity approx. 3398 kJ/h |
| Heat Value | 3398 kJ/h |
| Height (Metric) | 200 mm |
| Includes | Needle Valve, Safety Valve, and Automatic Gas shut-off |