Learn More
Gallenkamp Vacuum Oven with Digital Electronic PID Control
Product Code. 13025703
Change view
Click to view available options
Capacity (Metric):
31 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13025703 31 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13025703 Supplier Gallenkamp Supplier No. OVA031.XX3.5
  • ONLINE EXCLUSIVE PRICE
    5065.00€
    4950.21€ / Pack of 1
    Save 114.79€
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-01-2027
Gallenkamp Vacuum Oven is insulated with special grade glass wool to reduce the radiation heat loss to the minimum.
  • Double walled chamber, outer chamber rectangular in shape made of thick mild steel sheets
  • The inner chamber is cylindrical in shape made of stainless steel sheets with provision for vacuum pump and the vacuum gauge is provided at the top of unit
  • The inner chamber accommodate one no. Stainless steel tray at adjustable height.
  • The oven is insulated with special grade glass wool to reduce the radiation heat loss to the minimum
  • The door is of stainless steel plate with a specially designed Mechanism having alignment
  • A perfect seal is obtained by a positive screw specially designed for tightening the lid
  • A glass window is provided in the centre for viewing without opening the door
  • A set of band heaters is wrapped around the vacuum chamber for fast response and maximum uniformity of heat
Specifications
Capacity (Metric) 31 L
Temperature (Metric) 200°C
Type Vacuum Oven
Material Stainless Steel