Learn More
Texwipe™ TechniCloth™ Mop Cover
Product Code. p-7161491 Shop All Texwipe Products
Change view
Click to view available options
Dimensions (L x W):
37.5 x 67.5 cm
59 x 25 cm
Unit Size:
Pack of 150
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15732416 37.5 x 67.5 cm Pack of 150
15742416 59 x 25 cm Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15732416 Supplier Texwipe™ Supplier No. TX627C
Only null left
Add to basket
Add to basket
Fabricated from a hydroentangled, nonwoven blend of 55% cellulose and 45% polyester
  • Fabric blend combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic wiper
  • With its low particle counts, low extractable levels and solvent compatibility, TechniCloth™ is ideal for general wiping and spill control in controlled environments
  • Ideal for applications where high absorbency and low extractables are prime considerations
  • Excellent for picking up aqueous spills
  • Excellent wet and dry strength due to the polyester
  • Highly absorbent due to the cellulose
  • Compatible with most solvents
  • Contains no chemical binders
  • Low extractable levels
Specifications
Material Polyester Cellulose
Dimensions (L x W) 37.5 x 67.5 cm
Packaging Quantity 150 mop covers/Bag