Learn More
Teats for Pasteur pipettes
Product Code. p-7116185
Click to view available options
Color:
Clear
Red
Unit Size:
Pack of 100
Product Code. 15748007
Brand: MBL BPP002
For use with Soda-lime Pasteur pipettes.
  • For use with Soda-lime Pasteur pipettes
TRUSTED_SUSTAINABILITY
Specifications
Description Teats for Pasteur pipettes
Color Clear
Material Silicone
For Use With (Equipment) Soda-lime Pasteur pipettes
Quantity 100/Pk.