Learn More
Equip Labo™ TE Fan
Click to view available options
Width (Metric):
370 mm
Unit Size:
Each
Description
- CE TE Fan 1500T/M 370W 220V
- Ventilator for fume cupboards
- Centrifugal fan on the way or on the terrace
- Maximum external connection at 5 m linear
- Support chair and proximity switch included
- For extraction hoods, standard and laboratory fume cupboards
- Centrifugal fan on the way or on the terrace
Specifications
Specifications
| Product Type | TE Fan |
| Quantity | 1 Ea. |
| Width (Metric) | 370 mm |