Learn More
Equip Labo™ TE Fan
Product Code. 12548075
Change view
Click to view available options
Width (Metric):
370 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
12548075 370 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12548075 Supplier Equip Labo™ Supplier No. 5045
  • ONLINE EXCLUSIVE PRICE
    2550.00€
    2125.59€ / Pack of 1
    Save 424.41€
    16% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Robust and reliable connection accessory for casing fan.
    • CE TE Fan 1500T/M 370W 220V
    • Ventilator for fume cupboards
    • Centrifugal fan on the way or on the terrace
    • Maximum external connection at 5 m linear
    • Support chair and proximity switch included
    • For extraction hoods, standard and laboratory fume cupboards
    • Centrifugal fan on the way or on the terrace
Specifications
Product Type TE Fan
Quantity 1 Ea.
Width (Metric) 370 mm