Learn More
Equip Labo™ TE Fan
Product Code. 12636732
Change view
Click to view available options
Width: 370mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12636732 Width: 370mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12636732 Supplier Equip Labo™ Supplier No. 5044
  • ONLINE EXCLUSIVE PRICE
    5515.00€
    4615.08€ / Pack of 1
    Save 899.92€
    16% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Robust and reliable connection accessory for casing fan.
    • CE TE Fan 1500T/M 370W 220V
    • Ventilator for fume cupboards
    • Centrifugal fan on the way or on the terrace
    • Maximum external connection at 5 m linear
    • Support chair and proximity switch included
    • For extraction hoods, standard and laboratory fume cupboards
    • Centrifugal fan on the way or on the terrace
Specifications
Product Type TE Fan
Quantity 1 Ea.
Width (Metric) 370 mm