Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cassettes
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
TE 22 Mini Tank Transfer Unit
TE 42, TE 62 Transfer Tank
Type:
Gel Cassette
Hinged Cassette
Unit Size:
Each
Description
- Accessories and Replacement Parts for TE62
Specifications
Specifications
| For Use With (Equipment) | TE 42, TE 62 Transfer Tank |
| Type | Hinged Cassette |
| Includes | 1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick |