Description
- Tested for use with Amersham ECL Western blotting reagents
- Built-in power supply
- Use minimal buffer - only enough to saturate blotting papers and membranes
- Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers
- Use low current and voltage for transfers
Specifications
Specifications
| For Use With (Equipment) | Amersham ECL Semi Dry Blotter |
| Type | TE 77 PWR Semi Dry Transfer Unit |