missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TCS JNK 5a

Product Code. 18074082
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074082 50 mg 50mg
18034182 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074082 Supplier R&D Systems™ Supplier No. 2827/50
  • ONLINE EXCLUSIVE PRICE
    633.00€
    598.50€ / 50mg
    Save 34.50€
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of JNK2 and JNK3

Selective inhibitor of JNK2 and JNK3 (pIC50 values are 6.7, 6.5, <5.0 and <4.8 for JNK3, JNK2, JNK1 and p38α respectively). Displays no significant activity at a range of other protein kinases including EGFR, ErbB2, cdk2, PLK-1 and Src (pIC50 < 5.0).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312917-14-9
Molecular Formula C20H16N2OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 332.42
Purity >99%
Molecular Weight (g/mol) 332.421
Chemical Name or Material TCS JNK 5a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.