Learn More
TCEP Hydrochloride, TRC
Product Code. 30009478
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009478 1 g 1g
30004865 10 g 10g
30003306 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009478 Supplier Toronto Research Chemicals Supplier No. TRCT0125001G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51805-45-9
Molecular Formula C9 H15 O6 P . Cl H
Molecular Weight (g/mol) 286.65
Synonym Tris (2-Carboxyethyl) Phosphine Hydrochloride (TCEP)
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES Cl.OC(=O)CCP(CCC(=O)O)CCC(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H
Formula Weight 286.0373
Percent Purity >95%
Chemical Name or Material TCEP Hydrochloride
Grade HPLC